2-Mesitylenesulphonyl chloride ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2,4,6-Trimethylbenzenesulphonyl chloride, Mesitylene-2-sulphonyl chloride
A11775.14
A11775.22
A11775.36
A11775.14EA
1550
CZK
A11775.14
A11775.22
A11775.36
2-Mesitylenesulphonyl chloride ≥99%
2-Mesitylenesulphonyl chloride
Purity: 99%
CAS No.: 773-64-8
Molecular Formula: C9H11ClO2S
Formula Weight: 218.70
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₉H₁₁ClO₂S MW: 218.7 Bod varu: 150 °C (20 mmHg) Bod tání: 53…57 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007434 CAS číslo: 773-64-8 EINECS: 212-257-8 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...