2,3,5-Trimethylhydroquinone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B22819.22
B22819.36
B22819.22EA
1320
CZK
B22819.22
B22819.36
2,3,5-Trimethylhydroquinone ≥98%
2,3,5-Trimethylhydroquinone
Purity: 98%
CAS No.: 700-13-0
Molecular Formula: C9H12O2
Formula Weight: 152.20
Flash Point: 140°(284°F)
Melting Point: 169-174°
Beilstein Reference: 1909183
UN No.: 3077
MDL No.: MFCD00002346
Vzorec:
C₉H₁₂O₂ Bod varu: 295 °C (1013 hPa) Bod tání: 169…174 °C Bod vzplanutí: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002346 CAS číslo: 700-13-0 EINECS: 211-838-3 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...