Tisk…

2,5-Dimethylbenzyl alcohol ≥97%

Dodavatel: Thermo Fisher Scientific

L01624.03 L01624.06
L01624.03EA 1830 CZK
L01624.03 L01624.06
2,5-Dimethylbenzyl alcohol ≥97%
2,5-Dimethylbenzyl alcohol

Purity: 97+% CAS No.: 53957-33-8 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 107°(224°F) Refractive Index: 1.5360 Melting Point: 40-42° Boiling Point: 232-234° Beilstein Reference: 2243420 MDL No.: MFCD00004625

Vzorec: C₉H₁₂O
Bod varu: 232…234 °C
Bod tání: 40…42 °C
Hustota: 1,011
Bod vzplanutí: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004625
CAS číslo: 53957-33-8
EINECS: 258-888-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR