3,5-Dimethylbenzyl alcohol ≥99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
(3,5-Dimethylphenyl)methanol, 5-(Hydroxymethyl)-m-xylene, 3,5-Dimethylbenzylalcohol
L14726.03
L14726.06
L14726.14
L14726.03EA
1140
CZK
L14726.03
L14726.06
L14726.14
3,5-Dimethylbenzyl alcohol ≥99%
3,5-Dimethylbenzyl alcohol
Purity: 99%
CAS No.: 27129-87-9
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 106°(222°F)
Refractive Index: 1.5310
Boiling Point: 218-221°
Beilstein Reference: 2077285
MDL No.: MFCD00004648
Vzorec:
C₉H₁₂O MW: 136.19 Bod varu: 218…221 °C Bod vzplanutí: 106 °C (222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004648 CAS číslo: 27129-87-9 EINECS: 248-241-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...