Tisk…

2,4,6-Trimethylbenzoyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,4,6-Trimethylbenzene-1-carbonyl chloride, 2,4,6-Trimethylbenzoyl Chloride

M-Clarity™ quality level = MQ100

682519-1G 682519-5G
SIAL682519-1GEA 3290 CZK
SIAL682519-1G SIAL682519-5G
2,4,6-Trimethylbenzoyl chloride, Sigma-Aldrich®
2,4,6-Trimethylbenzoyl chloride
Vzorec: C₁₀H₁₁ClO
MW: 182,65 g/mol
Bod varu: 48 °C (0,05 mmHg)
Hustota: 1,0967 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00013650
CAS číslo: 938-18-1
EINECS: 213-339-6
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR