2,4,6-Triisopropylbenzoyl chloride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A11280.06
A11280.14
A11280.22
A11280.06EA
2850
CZK
A11280.06
A11280.14
A11280.22
2,4,6-Triisopropylbenzoyl chloride ≥98%
2,4,6-Triisopropylbenzoyl chloride
Purity: 98+%
CAS No.: 57199-00-5
Molecular Formula: C16H23ClO
Formula Weight: 266.81
Melting Point: 78-82°
Beilstein Reference: 2269534
UN No.: 3261
MDL No.: MFCD00015030
Vzorec:
C₁₆H₂₃ClO MW: 266,81 g/mol Bod varu: 107…108 °C (2 mmHg) Bod tání: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015030 CAS číslo: 57199-00-5 EINECS: 260-614-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...