Tisk…

2',4'-Dimethylacetophenone ≥97%

Dodavatel: Thermo Fisher Scientific

A12992.18 A12992.30
A12992.18EA 3780 CZK
A12992.18 A12992.30
2',4'-Dimethylacetophenone ≥97%
2',4'-Dimethylacetophenone

Purity: 97% CAS No.: 89-74-7 Molecular Formula: C10H12O Formula Weight: 148.21 Flash Point: 100°(212°F) Refractive Index: 1.5350 Odour: Characteristic Boiling Point: 227-228° Beilstein Reference: 956772 MDL No.: MFCD00003571

Vzorec: C₁₀H₁₂O
Bod varu: 227…228 °C
Hustota: 0,999
Bod vzplanutí: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003571
CAS číslo: 89-74-7
EINECS: 201-935-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR