4,4'-Dimethylbiphenyl ≥99%
Dodavatel: Thermo Fisher Scientific
B25089.03
B25089.06
B25089.14
B25089.03EA
1390
CZK
B25089.03
B25089.06
B25089.14
4,4'-Dimethylbiphenyl ≥99%
4,4'-Dimethylbiphenyl
Purity: 99%
CAS No.: 613-33-2
Molecular Formula: C14H14
Formula Weight: 182.27
Melting Point: 118-122°
Boiling Point: 295°
MDL No.: MFCD00008545
Vzorec:
C₁₄H₁₄ Bod varu: 295 °C Bod tání: 118…122 °C Hustota: 1,102 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008545 CAS číslo: 613-33-2 EINECS: 210-337-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...