4-Nitrobenzophenone ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
p-Nitrobenzophenone
B20405.06
B20405.14
B20405.06EA
628
CZK
B20405.06
B20405.14
4-Nitrobenzophenone ≥99%
4-Nitrobenzophenone
Purity: 99%
CAS No.: 1144-74-7
Molecular Formula: C13H9NO3
Formula Weight: 227.22
Melting Point: 136-138°
Beilstein Reference: 1912753
Vzorec:
C₁₃H₉NO₃ MW: 227,22 g/mol Bod tání: 136…138 °C Hustota: 1,406 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007354 CAS číslo: 1144-74-7 EINECS: 214-542-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...