4,4'-Bis(diethylamino)benzophenone, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone, 4,4-Bis(diethylamino) benzophenone
M-Clarity™ quality level = MQ100
160326-25G
160326-100G
SIAL160326-25GEA
4580
CZK
SIAL160326-25G
SIAL160326-100G
4,4'-Bis(diethylamino)benzophenone, Sigma-Aldrich®
4,4'-Bis(diethylamino)benzophenone
Vzorec:
C₂₁H₂₈N₂O Bod varu: 420 °C (1013 hPa) Storage Temperature: Ambient |
MDL Number:
MFCD00009044 CAS číslo: 90-93-7 EINECS: 202-025-4 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...