p-Cymen ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1-Isopropyl-4-methylbenzene, 4-Methylcumene, 4-Isopropyltoluene
A19226.0F
A19226.AP
A19226.0FEA
2930
CZK
A19226.0F
A19226.AP
p-Cymen ≥97%
p-Cymen
Purity: 97%
CAS No.: 99-87-6
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 47°(116°F)
Refractive Index: 1.4900
Odour: Aromatic
Melting Point: -68°
Boiling Point: 176-178°
Beilstein Reference: 1903377
Merck Reference: 14,2763
UN No.: 2046
MDL No.: MFCD00008893
Vzorec:
C₁₀H₁₄ Bod varu: 176…178 °C Bod tání: –68 °C Hustota: 0,860 Bod vzplanutí: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008893 CAS číslo: 99-87-6 EINECS: 202-796-7 UN: 2046 ADR: 3,III Merck Index: 14,2763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...