Tisk…

O-Cymene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

255270-100MG 255270-500MG
SIAL255270-100MGEA 3330 CZK
SIAL255270-100MG SIAL255270-500MG
O-Cymene, Sigma-Aldrich®
O-Cymene
Vzorec: C₁₀H₁₄
Bod varu: 177,2 °C (1013 hPa)
Hustota: 0,8902 g/cm³ (0 °C)
MDL Number: MFCD00008888
CAS číslo: 527-84-4
EINECS: 208-426-0
UN: 2046
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR