m-Ethyltoluene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
3-Ethyltoluene, 1-Ethyl-3-methylbenzene
M-Clarity™ quality level = MQ100
E49606-10G
E49606-25G
SIALE49606-10GEA
5530
CZK
SIALE49606-10G
SIALE49606-25G
m-Ethyltoluene, Sigma-Aldrich®
m-Ethyltoluene
Vzorec:
C₉H₁₂ MW: 120.19 Bod varu: 158…160 °C (1013 hPa) Bod tání: –95.5 °C Hustota: 0.869 Storage Temperature: Ambient |
MDL Number:
MFCD00009259 CAS číslo: 620-14-4 EINECS: 210-626-8 UN: 3295 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...