Tisk…

4-(Hydroxymethyl)benzyl alcohol 99%

Dodavatel: Thermo Fisher Scientific

Synonyms: p-Phenylenedimethanol, 1,4-Benzenedimethanol, 1,4-Bis(hydroxymethyl)-benzene, p-Xylene-α,α'-diol

105280500 105280100
ACRO105280500EA 9930 CZK
ACRO105280500 ACRO105280100
4-(Hydroxymethyl)benzyl alcohol 99%
4-(Hydroxymethyl)benzyl alcohol
Vzorec: C₈H₁₀O₂
MW: 138.17
Bod varu: 138…143 °C (1 mmHg)
Bod tání: 116…119 °C
MDL Number: MFCD00004665
CAS číslo: 589-29-7
EINECS: 209-641-2

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 116 to 119 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR