Tisk…

2-(Hydroxymethyl)benzyl alcohol ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: o-Phenylenedimethanol, o-Xylene-α,α'-diol, 1,2-Benzenedimethanol

B23008.03 B23008.06 B23008.14
B23008.03EA 1460 CZK
B23008.03 B23008.06 B23008.14
2-(Hydroxymethyl)benzyl alcohol ≥97%
2-(Hydroxymethyl)benzyl alcohol

Purity: 97% CAS No.: 612-14-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 59-64° Beilstein Reference: 508775

Vzorec: C₈H₁₀O₂
MW: 138.17
Bod varu: 291
Bod tání: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00004626
CAS číslo: 612-14-6
EINECS: 210-293-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR