2,5-Xylenol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2,5-Dimethylphenol, 2-Hydroxy-p-xylene, 2-Hydroxy-4-xylene
M-Clarity™ quality level = MQ300
W359505-1KG-K
W359505-5KG-K
SIALW359505-1KG-KEA
6280
CZK
SIALW359505-1KG-K
SIALW359505-5KG-K
2,5-Xylenol, Sigma-Aldrich®
2,5-Xylenol
Vzorec:
C₈H₁₀O MW: 122.17 Bod varu: 212 Bod tání: 75 Hustota: 1.169 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002237 CAS číslo: 95-87-4 EINECS: 202-461-5 UN: 2261 ADR: 6.1 II Merck Index: 12,10215 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...