Tisk…

2,6-Dimethylanisole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

D146404-100G D146404-25G
SIALD146404-100GEA 6070 CZK
SIALD146404-100G SIALD146404-25G
2,6-Dimethylanisole, Sigma-Aldrich®
2,6-Dimethylanisole
Vzorec: C₉H₁₂O
MW: 136,19 g/mol
Bod varu: 182 °C (1013 hPa)
Hustota: 0,96 g/cm³ (20 °C)
MDL Number: MFCD00008380
CAS číslo: 1004-66-6
EINECS: 213-723-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR