Tisk…

3,5-Xylidine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,5-Dimethylaniline, 5-Amino-m-xylene

M-Clarity™ quality level = MQ200

137863-100ML 137863-500ML
SIAL137863-100MLEA 1490 CZK
SIAL137863-100ML SIAL137863-500ML
3,5-Xylidine, Sigma-Aldrich®
3,5-Xylidine
Vzorec: C₈H₁₁N
MW: 121.18
Bod varu: 220
Bod tání: 9.8
Hustota: 0.9706 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007813
CAS číslo: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1 II
Merck Index: 12,10217

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR