2,3-Dimethylaniline, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2,3-Xylidine, 3-Amino-o-xylene
M-Clarity™ quality level = MQ200
D145807-100G
D145807-500G
SIALD145807-100GEA
1810
CZK
SIALD145807-100G
SIALD145807-500G
2,3-Dimethylaniline, Sigma-Aldrich®
2,3-Dimethylaniline
Vzorec:
C₈H₁₁N MW: 121,18 g/mol Bod varu: 222 °C (1013 hPa) Bod tání: –15 °C Hustota: 0,99 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007732 CAS číslo: 87-59-2 EINECS: 201-755-0 UN: 1711 ADR: 6.1,II Merck Index: 12,10217 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...