4-Methylbenzylamine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
4-(Aminomethyl)toluene
M-Clarity™ quality level = MQ200
M31201-100G
M31201-25G
SIALM31201-100GEA
8720
CZK
SIALM31201-100G
SIALM31201-25G
4-Methylbenzylamine, Sigma-Aldrich®
4-Methylbenzylamine
Vzorec:
C₈H₁₁N MW: 121,18 g/mol Bod varu: 195 °C (1013 hPa) Bod tání: 20…21 °C Hustota: 0,952 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008123 CAS číslo: 104-84-7 EINECS: 203-243-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...