4-Methylbenzylamine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-(Aminomethyl)toluene
A13768.14
A13768.22
A13768.14EA
1950
CZK
A13768.14
A13768.22
4-Methylbenzylamine 98%
4-Methylbenzylamine
Purity: 98%
CAS No.: 104-84-7
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 75°(167°F)
Refractive Index: 1.5350
Melting Point: 12-13°
Boiling Point: 196-197°
Beilstein Reference: 956670
UN No.: 2735
MDL No.: MFCD00008123
Vzorec:
C₈H₁₁N MW: 121,18 g/mol Bod varu: 196…197 °C Bod tání: 12…13 °C Hustota: 0,955 Bod vzplanutí: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008123 CAS číslo: 104-84-7 EINECS: 203-243-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...