Tisk…

2-Methylbenzylamine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: o-Tolylmethanamine, 2-(Aminomethyl)toluene

L05243.03 L05243.06 L05243.14
L05243.03EA 1090 CZK
L05243.03 L05243.06 L05243.14
2-Methylbenzylamine 98%
2-Methylbenzylamine

Purity: 98% CAS No.: 89-93-0 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 83°(181°F) Refractive Index: 1.5440 Boiling Point: 198-200° Beilstein Reference: 1099406 UN No.: 2735 MDL No.: MFCD00008112

Vzorec: C₈H₁₁N
MW: 121,18 g/mol
Bod varu: 198…200 °C
Bod tání: < 0 °C
Hustota: 0,979
Bod vzplanutí: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008112
CAS číslo: 89-93-0
EINECS: 201-952-1
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR