3,5-Dimethylphenyl isocyanate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3,5-Xylyl isocyanate, 3,5-Dimethylphenylisocyanate
390100-25G
390100-5G
SIAL390100-25GEA
14230
CZK
SIAL390100-25G
SIAL390100-5G
3,5-Dimethylphenyl isocyanate, Sigma-Aldrich®
3,5-Dimethylphenyl isocyanate
Vzorec:
C₉H₉NO MW: 147,18 g/mol Bod varu: 205 °C (1013 hPa) Hustota: 1,04 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013868 CAS číslo: 54132-75-1 EINECS: 258-987-0 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...