2,4-Dimethylphenyl isocyanate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2,4-Xylyl isocyanate, 1-Isocyanato-2,4-dimethylbenzene
L11126.03
L11126.06
L11126.03EA
944
CZK
L11126.03
L11126.06
2,4-Dimethylphenyl isocyanate ≥98%
2,4-Dimethylphenyl isocyanate
Purity: 98+%
CAS No.: 51163-29-2
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 87°(188°F)
Refractive Index: 1.5350
Boiling Point: 114-115°/2mm
Beilstein Reference: 386756
UN No.: 2206
MDL No.: MFCD00013853
Vzorec:
C₉H₉NO Bod varu: 114…115 °C (2 mmHg) Hustota: 1,041 Bod vzplanutí: 87 °C (188 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013853 CAS číslo: 51163-29-2 UN: 2206 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...