Tisk…

2,6-Diisopropylphenyl isocyanate ≥94%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2,6-Diisopropylphenylisocyanate

L10613.06 L10613.14 L10613.03
L10613.06EA 2580 CZK
L10613.06 L10613.14 L10613.03
2,6-Diisopropylphenyl isocyanate ≥94%
2,6-Diisopropylphenyl isocyanate

Purity: 94% CAS No.: 28178-42-9 Molecular Formula: C13H17NO Formula Weight: 203.29 Storage Temperature: 2°C to 8°C Flash Point: 107°(224°F) Refractive Index: 1.5180 Boiling Point: 116-117°/10mm Beilstein Reference: 2645721 UN No.: 2206 MDL No.: MFCD00008882

Vzorec: C₁₃H₁₇NO
MW: 203,28 g/mol
Bod varu: 116…117 °C (10 mmHg)
Hustota: 0,951
Bod vzplanutí: 107 °C (224 °F)
MDL Number: MFCD00008882
CAS číslo: 28178-42-9
EINECS: 248-885-4
UN: 2206
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR