Tisk…

5-Iodo-m-xylene 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Iodo-3,5-dimethylbenzene, 3,5-Dimethyl-1-iodobenzene, 1,3-Dimethyl-5-iodobenzene, 3,5-Dimethyliodobenzene

A18413.09 A18413.18
A18413.09EA 1440 CZK
A18413.09 A18413.18
5-Iodo-m-xylene 97%
5-Iodo-m-xylene

Purity: 97% CAS No.: 22445-41-6 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 110°(230°F) Refractive Index: 1.5950 Boiling Point: 229° Beilstein Reference: 2039353 MDL No.: MFCD00060659

Vzorec: C₈H₉I
MW: 232,06 g/mol
Bod varu: 229 °C
Hustota: 1,610
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060659
CAS číslo: 22445-41-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR