2-Iodo-m-xylene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1,3-Dimethyl-2-iodobenzene, 2-Iodo-1,3-dimethylbenzene, 2,6-Dimethyliodobenzene, 2,6-Dimethyl-1-iodobenzene
A13439.06
A13439.14
A13439.06EA
2710
CZK
A13439.06
A13439.14
2-Iodo-m-xylene ≥98%
2-Iodo-m-xylene
Purity: 98%
CAS No.: 608-28-6
Molecular Formula: C8H9I
Formula Weight: 232.07
Flash Point: 110°(230°F)
Refractive Index: 1.6025
Boiling Point: 230-231°
Beilstein Reference: 1857414
MDL No.: MFCD00019012
Vzorec:
C₈H₉I Bod varu: 230…231 °C Hustota: 1,620 Bod vzplanutí: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019012 CAS číslo: 608-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...