Tisk…

3-Iodo-o-xylene ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1,2-Dimethyl-3-iodobenzene, 2,3-Dimethyl-1-iodobenzene, 1-Iodo-2,3-dimethylbenzene, 2,3-Dimethyliodobenzene

A11774.06 A11774.14
A11774.06EA 1430 CZK
A11774.06 A11774.14
3-Iodo-o-xylene ≥97%
3-Iodo-o-xylene

Purity: 97% CAS No.: 31599-60-7 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 110°(230°F) Refractive Index: 1.6030 Boiling Point: 98-100°/15mm Beilstein Reference: 1929733 MDL No.: MFCD00041437

Vzorec: C₈H₉I
MW: 232,06 g/mol
Bod varu: 98…100 °C (15 mmHg)
Hustota: 1,640
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041437
CAS číslo: 31599-60-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR