Tisk…

α,α,α,α',α',α'-Hexafluoro-3,5-xylidine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline

A15289.09 A15289.18
A15289.09EA 817 CZK
A15289.09 A15289.18
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine ≥98%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine

Purity: 98% CAS No.: 328-74-5 Molecular Formula: C8H5F6N Formula Weight: 229.13 Flash Point: 83°(181°F) Refractive Index: 1.4340 Odour: Recognizable Boiling Point: 75-78°/10mm Beilstein Reference: 654318 UN No.: 2810 MDL No.: MFCD00000394

Vzorec: C₈H₅F₆N
Bod varu: 75…78 °C (10 mmHg)
Bod tání: 3 °C
Hustota: 1,473
Bod vzplanutí: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS číslo: 328-74-5
EINECS: 206-335-0
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR