3,5-Dimethyl-p-anisic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3,5-Dimethyl-p-anisic acid, 4-Methoxy-3,5-dimethylbenzoic acid, 3,5-Dimethyl-4-methoxybenzenecarboxylic acid, 3,5-Dimethyl-4-methoxybenzoic acid
M-Clarity™ quality level = MQ100
638692-1G
638692-5G
SIAL638692-1GEA
0
CZK
SIAL638692-1G
SIAL638692-5G
3,5-Dimethyl-p-anisic acid, Sigma-Aldrich®
3,5-Dimethyl-p-anisic acid
Vzorec:
C₁₀H₁₂O₃ MW: 180,2 g/mol Bod varu: 313 °C (1013 hPa) Bod tání: 196…198 °C Hustota: 1,253 g/cm³ (20 °C) |
MDL Number:
MFCD00020309 CAS číslo: 21553-46-8 EINECS: 244-441-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...