Tisk…

3'-Methylacetophenone, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Methyl m-tolyl ketone, 1-(m-Tolyl)ethanone, m-Methylacetophenone, 3′-Methylacetophenone

M-Clarity™ quality level = MQ200

M26607-25G M26607-5G
SIALM26607-25GEA 3530 CZK
SIALM26607-25G SIALM26607-5G
3'-Methylacetophenone, Sigma-Aldrich®
3'-Methylacetophenone
Vzorec: C₉H₁₀O
MW: 134.18
Storage Temperature: Ambient
MDL Number: MFCD00008742
CAS číslo: 585-74-0
EINECS: 209-561-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR