α,α'-Dibromo-p-xylene ≥97%, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
p-Xylylene dibromide, 1,4-Bis(bromomethyl)benzene
M-Clarity™ quality level = MQ100
D44804-25G
D44804-5G
SIALD44804-25GEA
2780
CZK
SIALD44804-25G
SIALD44804-5G
α,α'-Dibromo-p-xylene ≥97%, Sigma-Aldrich®
α,α'-Dibromo-p-xylene
Vzorec:
C₈H₈Br₂ MW: 263.96 Bod varu: 245 °C (lit.) Bod tání: 143…145 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00000182 CAS číslo: 623-24-5 EINECS: 210-781-1 UN: 3448 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...