α,α'-Dichloro-p-xylene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,4-Bis(chloromethyl)benzene, p-Xylene dichloride, p-Xylylene dichloride
M-Clarity™ quality level = MQ100
105740-100G
105740-25G
SIAL105740-100GEA
2630
CZK
SIAL105740-100G
SIAL105740-25G
α,α'-Dichloro-p-xylene, Sigma-Aldrich®
α,α'-Dichloro-p-xylene
Vzorec:
C₈H₈Cl₂ Bod varu: 254 °C (1013 hPa) Bod tání: 100 °C Hustota: 1,432 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000920 CAS číslo: 623-25-6 EINECS: 210-782-7 UN: 2928 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...