α,α'-Dichloro-o-xylene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
o-Xylylene Dichloride, o-Xylylene chloride, α,α′-Dichloro-o-xylene, α,α'-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, Alpha,Alpha’-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, o-Xylene dichloride
A12500.18
A12500.30
A12500.18EA
1860
CZK
A12500.18
A12500.30
α,α'-Dichloro-o-xylene ≥98%
α,α'-Dichloro-o-xylene
Purity: 98%
CAS No.: 612-12-4
Molecular Formula: C8H8Cl2
Formula Weight: 175.06
Flash Point: 107°(224°F)
Odour: Pungent
Boiling Point: 246°
Beilstein Reference: 2043675
UN No.: 2928
MDL No.: MFCD00000903
Vzorec:
C₈H₈Cl₂ MW: 175.06 Bod varu: 246 °C Bod tání: 53…57 °C Hustota: 1.393 g/cm³ (0 °C) Bod vzplanutí: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000903 CAS číslo: 612-12-4 EINECS: 210-291-8 UN: 2928 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...