2,5-Dimethylbenzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
151068-25G
151068-5G
SIAL151068-25GEA
10240
CZK
SIAL151068-25G
SIAL151068-5G
2,5-Dimethylbenzaldehyde, Sigma-Aldrich®
2,5-Dimethylbenzaldehyde
Vzorec:
C₉H₁₀O MW: 134,18 g/mol Bod varu: 104,4 °C (1013 hPa) Hustota: 0,95 g/cm³ (20 °C) |
MDL Number:
MFCD00003342 CAS číslo: 5779-94-2 EINECS: 227-303-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...