2-Bromo-4,6-xylidine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
6-Bromo-2,4-dimethylaniline, 6-Bromo-2,4-xylidine, 2-Bromo-4,6-dimethylaniline
B21272.09
B21272.14
B21272.18
B21272.09EA
2230
CZK
B21272.09
B21272.14
B21272.18
2-Bromo-4,6-xylidine ≥98%
2-Bromo-4,6-xylidine
Purity: 98%
CAS No.: 41825-73-4
Molecular Formula: C8H10BrN
Formula Weight: 200.08
Flash Point:
Vzorec:
C₈H₁₀BrN Bod varu: 75…79 °C (5 mmHg) Bod tání: 42…46 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00047826 CAS číslo: 41825-73-4 EINECS: 246-337-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...