α'-Bromo-α,α,α-trifluoro-p-xylene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
4-(Bromomethyl)benzotrifluoride, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)benzyl bromide
M-Clarity™ quality level = MQ100
290564-1G
290564-25G
290564-5G
SIAL290564-1GEA
1210
CZK
SIAL290564-1G
SIAL290564-25G
SIAL290564-5G
α'-Bromo-α,α,α-trifluoro-p-xylene, Sigma-Aldrich®
α'-Bromo-α,α,α-trifluoro-p-xylene
Vzorec:
C₈H₆BrF₃ MW: 239.03 Bod varu: 73…75 °C (0.4 mmHg) Bod tání: 31.5 Hustota: 1.54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000403 CAS číslo: 402-49-3 EINECS: 206-947-8 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...