Tisk…

α'-Bromo-α,α,α-trifluoro-m-xylene, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 3-(Trifluoromethyl)benzyl bromide, 1-(Bromomethyl)-3-(trifluoromethyl)benzene, 3-(Bromomethyl)benzotrifluoride

M-Clarity™ quality level = MQ200

290556-25G 290556-5G
SIAL290556-25GEA 7070 CZK
SIAL290556-25G SIAL290556-5G
α'-Bromo-α,α,α-trifluoro-m-xylene, Sigma-Aldrich®
α'-Bromo-α,α,α-trifluoro-m-xylene
Vzorec: C₈H₆BrF₃
MW: 239.03
Bod varu: 50 °C (0.3 mmHg)
Hustota: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00000395
CAS číslo: 402-23-3
EINECS: 206-938-9
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR