Tisk…

α,α'-Dibromo-m-xylene, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide

M-Clarity™ quality level = MQ200

125911-10G 125911-25G
SIAL125911-10GEA 4590 CZK
SIAL125911-10G SIAL125911-25G
α,α'-Dibromo-m-xylene, Sigma-Aldrich®
α,α'-Dibromo-m-xylene
Vzorec: C₈H₈Br₂
Bod varu: 135…140 °C (20 mmHg)
Bod tání: 76…78 °C
Hustota: 1,959 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000178
CAS číslo: 626-15-3
EINECS: 210-931-6
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR