3,5-Dimethylphenylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B23434.03
B23434.06
B23434.14
B23434.03EA
1220
CZK
B23434.03
B23434.06
B23434.14
3,5-Dimethylphenylboronic acid ≥98%
3,5-Dimethylphenylboronic acid
Purity: 98%
CAS No.: 172975-69-8
Molecular Formula: C8H11BO2
Formula Weight: 149.99
Melting Point: 235-237°
Beilstein Reference: 7368658
Vzorec:
C₈H₁₁BO₂ Bod tání: 235…237 °C Storage Temperature: Ambient |
MDL Number:
MFCD00185689 CAS číslo: 172975-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...