2,6-Dimethylphenylboronic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,6-Dimethylbenzeneboronic acid, (2,6-Dimethylphenyl)boronic acid, Dihydroxy(2,6-dimethylphenyl)borane, (2,6-Dimethylphenyl)metaboric acid
M-Clarity™ quality level = MQ100
480061-25G
480061-5G
SIAL480061-25GEA
14020
CZK
SIAL480061-25G
SIAL480061-5G
2,6-Dimethylphenylboronic acid, Sigma-Aldrich®
2,6-Dimethylphenylboronic acid
Vzorec:
C₈H₁₁BO₂ MW: 149,98 g/mol |
MDL Number:
MFCD01009693 CAS číslo: 100379-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...