Tisk…

2-Biphenylylmethanol ≥98%

Dodavatel: Thermo Fisher Scientific

L07094.03 L07094.06
L07094.03EA 900 CZK
L07094.03 L07094.06
2-Biphenylylmethanol ≥98%
2-Biphenylylmethanol

Purity: 98% CAS No.: 2928-43-0 Molecular Formula: C13H12O Formula Weight: 184.24 Melting Point: 47-51° Boiling Point: 96°/0.04mm MDL No.: MFCD00004620

Vzorec: C₁₃H₁₂O
Bod varu: 96 °C (0,04 mmHg)
Bod tání: 47…51 °C
Hustota: 1,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004620
CAS číslo: 2928-43-0
EINECS: 220-892-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR