Tisk…

Benzhydryl alcohol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diphenylcarbinol, Benzhydrol, Diphenylmethanol

M-Clarity™ quality level = MQ100

B4856-100G B4856-500G B4856-5G
SIALB4856-100GEA 1150 CZK
SIALB4856-100G SIALB4856-500G SIALB4856-5G
Benzhydryl alcohol, Sigma-Aldrich®
Benzhydryl alcohol
Vzorec: C₁₃H₁₂O
MW: 184,24 g/mol
Bod varu: 297 °C (1013 hPa)
Bod tání: 63…64 °C
Hustota: 1,054 g/cm³ (80 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004488
CAS číslo: 91-01-0
EINECS: 202-033-8
Merck Index: 12,01121

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR