Tisk…

Trityl alcohol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Triphenylmethanol, Triphenylcarbinol

M-Clarity™ quality level = MQ200

134848-250G 134848-50G
SIAL134848-250GEA 5490 CZK
SIAL134848-250G SIAL134848-50G
Trityl alcohol, Sigma-Aldrich®
Trityl alcohol
Vzorec: C₁₉H₁₆O
MW: 260.34
Bod varu: 360
Bod tání: 160…163 °C
Hustota: 1.1884 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004445
CAS číslo: 76-84-6
EINECS: 200-988-5
Merck Index: 13,09810

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR