Tisk…

Trityl alcohol ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Triphenylmethanol, Triphenylcarbinol

A10366.0B A10366.18 A10366.30
A10366.0BEA 13390 CZK
A10366.0B A10366.18 A10366.30
Trityl alcohol ≥98%
Trityl alcohol

Purity: 98% CAS No.: 76-84-6 Molecular Formula: (C6H5)3COH Formula Weight: 260.34 Physical Form: Cryst powder Appearance: White to cream Melting Point: 161-164° Boiling Point: 360-380° Beilstein Reference: 1460837 Merck Reference: 14,9739 MDL No.: MFCD00004445

Vzorec: C₁₉H₁₆O
MW: 260,34 g/mol
Bod varu: 360…380 °C
Bod tání: 161…164 °C
Hustota: 1,1884 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004445
CAS číslo: 76-84-6
EINECS: 200-988-5
Merck Index: 14,9739

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR