Tisk…

Dibenzyl ether, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Benzyl ether

M-Clarity™ quality level = MQ300

W237108-10KG-K W237108-1KG-K
SIALW237108-10KG-KEA 12550 CZK
SIALW237108-10KG-K SIALW237108-1KG-K
Dibenzyl ether, Sigma-Aldrich®
Dibenzyl ether
Vzorec: C₆H₅CH₂OCH₂C₆H₅
MW: 198,26 g/mol
Bod varu: 298 °C (1013 hPa)
Hustota: 1,0424 g/cm³ (20 °C)
MDL Number: MFCD00004780
CAS číslo: 103-50-4
EINECS: 203-118-2
UN: 3082
ADR: 9,III
Merck Index: 13,01134
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cb077d69cfe8c3315f62058a54ca8460d752735&vid=08fffe90d8cf2c6ddb596093c7e9631247e2a70b&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR