Dibenzyl ether ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
|
Synonyms:
Benzyl ether
A18447.0E
A18447.30
A18447.36
A18447.0EEA
2790
CZK
A18447.0E
A18447.30
A18447.36
Dibenzyl ether ≥98%
Dibenzyl ether
Purity: 98+%
CAS No.: 103-50-4
Molecular Formula: C6H5CH2OCH2C6H5
Formula Weight: 198.27
Flash Point: 135°(275°F)
Refractive Index: 1.5620
Melting Point: 3-4°
Boiling Point: 298°
Beilstein Reference: 1911156
Merck Reference: 14,1132
UN No.: 3082
MDL No.: MFCD00004780
Vzorec:
C₆H₅CH₂OCH₂C₆H₅ MW: 198.26 Bod varu: 298 °C Bod tání: 3…4 °C Hustota: 1.040 Bod vzplanutí: 135 °C (275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004780 CAS číslo: 103-50-4 EINECS: 203-118-2 UN: 3082 ADR: 9 III Merck Index: 13,01134 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...