Tisk…

Dibenzylamine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A11554.0B A11554.30
A11554.0BEA 4570 CZK
A11554.0B A11554.30
Dibenzylamine 98%
Dibenzylamine

Purity: 98% CAS No.: 103-49-1 Molecular Formula: C14H15N Formula Weight: 197.28 Flash Point: 143°(289°F) Refractive Index: 1.5745 Odour: Weak pH: 8.9 Melting Point: -26° Boiling Point: 300° Beilstein Reference: 909664 Merck Reference: 14,3011 UN No.: 2735 MDL No.: MFCD00004770

Vzorec: C₆H₅CH₂NHCH₂C₆H₅
MW: 197.28
Bod varu: 300 °C
Bod tání: –26 °C
Hustota: 1.026
Bod vzplanutí: 143 °C (289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004770
CAS číslo: 103-49-1
EINECS: 203-117-7
UN: 2735
ADR: 8 III
Merck Index: 13,03035
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c17e97ec30680cc9e622a04bff55a48e1b301831&vid=c74bd48fc42a0747a1603d963976594b20a27385&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR