2-Fluorobenzophenone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
o-Fluorobenzophenone
L09316.06
L09316.14
L09316.06EA
1570
CZK
L09316.06
L09316.14
2-Fluorobenzophenone ≥98%
2-Fluorobenzophenone
Purity: 98+%
CAS No.: 342-24-5
Molecular Formula: C13H9FO
Formula Weight: 200.21
Flash Point:
Vzorec:
C₁₃H₉FO MW: 200,21 g/mol Bod varu: 149…150 °C (16 mmHg) Hustota: 1,186 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000318 CAS číslo: 342-24-5 EINECS: 206-440-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...