Tisk…

Fenolftalein ACS, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

M-Clarity™ quality level = MQ200

105945-2.5KG 105945-4X2.5KG 105945-500G
SIAL105945-2.5KGEA 13500 CZK
SIAL105945-2.5KG SIAL105945-4X2.5KG SIAL105945-500G
Fenolftalein ACS, Sigma-Aldrich®
Fenolftalein
Vzorec: C₂₀H₁₄O₄
MW: 318.33
Bod tání: 258…262 °C
Hustota: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS číslo: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR